4,4,4-Trifluorocrotonic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 4,4,4-Trifluorobut-2-enoic acid

003694-1G 003694-5G 003694-25G
101777-036EA 21.54 USD
101777-036 101777-038 101777-040
4,4,4-Trifluorocrotonic acid ≥97%
4,4,4-Trifluorocrotonic acid

Hazard: Corrosive/Irritant/Refrigerate

Formula: C₄H₃F₃O₂
MW: 140.06 g/mol
Boiling Pt: 145…150 °C
Melting Pt: 54…56 °C
MDL Number: MFCD02181180
CAS Number: 71027-02-6

Order Now

Specification Test Results

Boiling point 145-150°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR