4,4,4-Trifluorobutyraldehyde hydrate, Technical Grade
Supplier: Matrix Scientific
Synonyms:
4,4,4-Trifluorobutanal hydrate
003687-5G
003687-1G
101777-028EA
892.31
USD
101777-028
101777-026
4,4,4-Trifluorobutyraldehyde hydrate, Technical Grade
4,4,4-Trifluorobutyraldehyde hydrate
Hazard: Flammable/Irritant/Refrigerate/Stored under Argon
Formula:
C₄H₇F₃O₂ MW: 144.09 g/mol Boiling Pt: 94-97°C |
CAS Number:
1448773-75-8 |
Specification Test Results
Boiling point | 94-97° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...