4,4,4-Trifluorobutyric acid ≥97%
Supplier: Matrix Scientific
003686-5G
003686-25G
003686-100G
101777-020EA
27.69
USD
101777-020
101777-022
101777-024
4,4,4-Trifluorobutyric acid ≥97%
4,4,4-Trifluorobutyric acid
Hazard: Corrosive/Irritant/Store in Freezer
Formula:
C₄H₅F₃O₂ MW: 142.08 g/mol Boiling Pt: 166-167°C Melting Pt: 30-31°C |
MDL Number:
MFCD00077604 CAS Number: 406-93-9 UN: 3261 ADR: 8,II |
Specification Test Results
Boiling point | 166-167° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...