1,2,3-Trifluorobenzene ≥98%

Supplier: Matrix Scientific

003682-25G 003682-100G
101777-008EA 95.38 USD
101777-008 101777-010
1,2,3-Trifluorobenzene ≥98%
1,2,3-Trifluorobenzene

Hazard: Flammable

Formula: C₆H₃F₃
Boiling Pt: 94-95°C
Density: 1.28
MDL Number: MFCD00012013
CAS Number: 1489-53-8
UN: 2387
ADR: 3,II

Order Now

Specification Test Results

Boiling point 94-95°
Density 1.28
ND20 1.423


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR