2,4,6-Trifluorobenzonitrile ≥98%

Supplier: Matrix Scientific

003681-25G 003681-5G
101777-006EA 98.46 USD
101777-006 101777-004
2,4,6-Trifluorobenzonitrile ≥98%
2,4,6-Trifluorobenzonitrile

Hazard: Toxic

Formula: C₇H₂F₃N
Boiling Pt: 179 °C
Melting Pt: 55…58 °C
MDL Number: MFCD00042399
CAS Number: 96606-37-0
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 92°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR