3-Bromo-4,5-diaminobenzotrifluoride ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Bromo-4,5-diamino-α,α,α-trifluorotoluene, 3-Bromo-5-(trifluoromethyl)-1,2-phenylenediamine, 1-Bromo-2,3-diamino-5-(trifluoromethyl)benzene, 3-Bromo-5-(trifluoromethyl)-o-phenylenediamine, 3-Bromo-5-(trifluoromethyl)benzene-1,2-diamine

001269-1G 001269-5G
101775-314EA 23.08 USD
101775-314 101775-316
3-Bromo-4,5-diaminobenzotrifluoride ≥97%
3-Bromo-4,5-diaminobenzotrifluoride

Hazard: Toxic

Formula: C₇H₆BrF₃N₂
MW: 255.04 g/mol
Melting Pt: 61-65°C
MDL Number: MFCD00042484
CAS Number: 113170-72-2
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 60°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR