Ethyl-4,4,4-trifluoroacetoacetate ≥99%
Supplier: Matrix Scientific
Synonyms:
Ethyl trifluoroacetoacetate, Ethyl 4,4,4-trifluoro-3-oxobutanoate
001255-250G
001255-1KG
101775-306EA
126.15
USD
101775-306
101775-308
Ethyl-4,4,4-trifluoroacetoacetate ≥99%
Ethyl-4,4,4-trifluoroacetoacetate
TCA listed: T
Hazard: Flammable
Formula:
C₆H₇F₃O₃ MW: 184.11 g/mol Boiling Pt: 129-130°C Density: 1.259 |
MDL Number:
MFCD00000424 CAS Number: 372-31-6 UN: 3272 ADR: 3,III |
Specification Test Results
Boiling point | 129-130° |
Density | 1.259 |
ND20 | 1.375 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...