Ethyl-4,4,4-trifluoro-3-(trifluoromethyl)crotonate ≥95%

Supplier: Matrix Scientific

001254-25G 001254-5G
101775-304EA 700 USD
101775-304 101775-302
Ethyl-4,4,4-trifluoro-3-(trifluoromethyl)crotonate ≥95%
Ethyl-4,4,4-trifluoro-3-(trifluoromethyl)crotonate

Hazard: Flammable/Moisture Sensitive/Stored under Argon

Formula: C₇H₆F₆O₂
MW: 236.12 g/mol
Boiling Pt: 128°C
MDL Number: MFCD00041517
CAS Number: 1513-60-6

Order Now

Specification Test Results

Boiling point 128°
ND20 1.3414

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR