Ethyl-4,4,4-trifluorobutyrate ≥98%

Supplier: Matrix Scientific

Synonyms: Ethyl 4,4,4-trifluorobutanoate, Ethyl 4,4,4-trifluorobutyrate

001253-5G 001253-25G
101775-296EA 53.85 USD
101775-296 101775-298
Ethyl-4,4,4-trifluorobutyrate ≥98%
Ethyl-4,4,4-trifluorobutyrate

Hazard: Flammable

Formula: C₆H₉F₃O₂
MW: 170.13 g/mol
Boiling Pt: 127 °C
Flash Pt: 33 °C
MDL Number: MFCD00041398
CAS Number: 371-26-6
UN: 3272
ADR: 3,III

Order Now

Specification Test Results

Boiling point 127°
ND20 1.352

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR