Ethyl-4,4,4-trifluoro-3-(trifluoromethyl)butyrate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl (2H-perfluoroprop-2-yl)acetate

001251-1G 001251-5G
101775-288EA 52.31 USD
101775-288 101775-290
Ethyl-4,4,4-trifluoro-3-(trifluoromethyl)butyrate ≥97%
Ethyl-4,4,4-trifluoro-3-(trifluoromethyl)butyrate

Hazard: Flammable

Formula: C₇H₈F₆O₂
MW: 238.13 g/mol
MDL Number: MFCD00236608
CAS Number: 17327-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR