5-Bromo-2,4-difluorobenzenesulfonyl chloride ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-2,4-difluorobenzene-1-sulphonyl chloride

001243-1G 001243-5G
101775-282EA 23.08 USD
101775-282 101775-284
5-Bromo-2,4-difluorobenzenesulfonyl chloride ≥95%
5-Bromo-2,4-difluorobenzenesulfonyl chloride

Hazard: Corrosive/Moisture Sensitive

Formula: C₆H₂BrClF₂O₂S
MW: 291.5 g/mol
Melting Pt: 34…36 °C
MDL Number: MFCD01940424
CAS Number: 287172-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR