2,2,3,4,4,4-Hexafluoro-1-butanol ≥95%
Supplier: Matrix Scientific
Synonyms:
1H,1H,3H-Perfluoro-1-butanol, 2,2,3,4,4,4-Hexafluorobutan-1-ol
001231-25G
001231-100G
101775-260EA
35.38
USD
101775-260
101775-262
2,2,3,4,4,4-Hexafluoro-1-butanol ≥95%
2,2,3,4,4,4-Hexafluoro-1-butanol
Hazard: Flammable
Formula:
C₄H₄F₆O MW: 182.07 g/mol Boiling Pt: 114 °C (740 mmHg) Density: 1.557 Flash Pt: 51 °C |
MDL Number:
MFCD00042309 CAS Number: 382-31-0 UN: 1987 ADR: 3 III |
Specification Test Results
Boiling point | 114°/740mm |
Density | 1.557 |
ND20 | 1.312 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...