1-Bromo-3,4,5-trifluorobenzene ≥99%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-1,2,3-trifluorobenzene

001225-25G 001225-100G
101775-252EA 23.08 USD
101775-252 101775-254
1-Bromo-3,4,5-trifluorobenzene ≥99%
1-Bromo-3,4,5-trifluorobenzene

Hazard: Flammable

Formula: C₆H₂BrF₃
MW: 210.98 g/mol
Boiling Pt: 47…49 °C (60 mmHg)
Density: 1.767
MDL Number: MFCD00070738
CAS Number: 138526-69-9
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Boiling point 47-49°/60m
Density 1.767
ND20 1.482

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR