2,2,3,3,4,4,4-Heptafluorobutylamine ≥98%

Supplier: Matrix Scientific

001209-25G 001209-5G
101775-244EA 915.38 USD
101775-244 101775-242
2,2,3,3,4,4,4-Heptafluorobutylamine ≥98%
2,2,3,3,4,4,4-Heptafluorobutylamine

Hazard: Toxic/Irritant-Harmful

Formula: C₄H₄F₇N
MW: 199.07 g/mol
Boiling Pt: 70-71°C
Density: 1.304
MDL Number: MFCD00014817
CAS Number: 374-99-2

Order Now

Specification Test Results

Boiling point 70-71°
Density 1.304
ND20 1.493

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR