1,1,1,2,2,3,3-Heptafluoro-5-iodopentane ≥97%

Supplier: Matrix Scientific

001166-1G 001166-5G
101775-226EA 38.46 USD
101775-226 101775-228
1,1,1,2,2,3,3-Heptafluoro-5-iodopentane ≥97%
1,1,1,2,2,3,3-Heptafluoro-5-iodopentane

Hazard: Irritant

Formula: C₅H₄F₇I
MW: 323.98 g/mol
Boiling Pt: 80 °C (200 mmHg)
MDL Number: MFCD00155834
CAS Number: 1513-88-8

Order Now

Specification Test Results

Boiling point 80°/200mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR