2,5-Bis(2,2,2-trifluoroethoxy)benzenesulfonylchloride ≥90%

Supplier: Matrix Scientific

001140-1G 001140-5G
101775-216EA 43.08 USD
101775-216 101775-218
2,5-Bis(2,2,2-trifluoroethoxy)benzenesulfonylchloride ≥90%
2,5-Bis(2,2,2-trifluoroethoxy)benzenesulfonylchloride

Hazard: Corrosive/Moisture Sensitive

Formula: C₁₀H₇ClF₆O₄S
MW: 372.67 g/mol
Melting Pt: 48…52 °C
MDL Number: MFCD00052841
CAS Number: 152457-95-9

Order Now

Specification Test Results

Melting point 48-52°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR