4-Bromo-1,1,2-trifluoro-1-butene ≥98%

Supplier: Matrix Scientific

001132-25G 001132-5G 001132-100G
101775-206EA 372.31 USD
101775-206 101775-204 101775-208
4-Bromo-1,1,2-trifluoro-1-butene ≥98%
4-Bromo-1,1,2-trifluoro-1-butene

Hazard: Flammable/Light Sensitive

Formula: C₄H₄BrF₃
MW: 188.97 g/mol
Boiling Pt: 98…100 °C
Density: 1.639
Flash Pt: 17 °C
MDL Number: MFCD00039274
CAS Number: 10493-44-4
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Boiling point 98-100°
ND20 1.4031

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR