2-Bromo-3',5'-bis(trifluoromethyl)acetophenone ≥95%
Supplier: Matrix Scientific
Synonyms:
1-(3,5-Bis(trifluoromethyl)phenyl)-2-bromoethanone, 3,5-Bis(trifluoromethyl)phenacyl bromide
001106-5G
001106-25G
101775-192EA
89.23
USD
101775-192
101775-194
2-Bromo-3',5'-bis(trifluoromethyl)acetophenone ≥95%
2-Bromo-3',5'-bis(trifluoromethyl)acetophenone
Hazard: Corrosive/Irritant/Refrigerate
Formula:
C₁₀H₅BrF₆O MW: 335.0405 g/mol Melting Pt: 49…51 °C |
MDL Number:
MFCD00792434 CAS Number: 131805-94-2 |
Specification Test Results
Melting point | 49-51° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...