Perfluoro(4-methyl-3,6-dioxaoct-7-ene)sulfonyl fluoride ≥95%

Supplier: Matrix Scientific

001082-5G 001082-25G
101775-188EA 180 USD
101775-188 101775-190
Perfluoro(4-methyl-3,6-dioxaoct-7-ene)sulfonyl fluoride ≥95%
Perfluoro(4-methyl-3,6-dioxaoct-7-ene)sulfonyl fluoride

TCA listed: T
Hazard: Irritant

Formula: C₇F₁₄O₄S
MW: 446.12 g/mol
Boiling Pt: 135 °C
Density: 1.7
Flash Pt: none
MDL Number: MFCD00798138
CAS Number: 16090-14-5

Order Now

Specification Test Results

Density 1.7

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR