ω-Bromo-2-fluoroacetophenone ≥97%

Supplier: Matrix Scientific

001067-5G 001067-25G 001067-1G
101775-184EA 67.69 USD
101775-184 101775-186 101775-182
ω-Bromo-2-fluoroacetophenone ≥97%
ω-Bromo-2-fluoroacetophenone

Hazard: Corrosive/Refrigerate/Lachrymator

Formula: C₈H₆BrFO
Boiling Pt: 239 °C
Melting Pt: 25…27 °C
Flash Pt: 98.6 °C
MDL Number: MFCD00278796
CAS Number: 655-15-2
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR