ω-Bromo-3,4-difluoroacetophenone ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-3',4'-difluoroacetophenone, α-Bromo-3,4-difluoroacetophenone, 2-Bromo-1-(3,4-difluorophenyl)ethan-1-one, 3,4-Difluorophenacyl bromide, 2-Bromo-1-(3,4-difluorophenyl)ethanone

001062-1G 001062-25G 001062-5G
101775-176EA 60 USD
101775-176 101775-180 101775-178
ω-Bromo-3,4-difluoroacetophenone ≥98%
ω-Bromo-3,4-difluoroacetophenone

Hazard: Corrosive

Formula: C₈H₅BrF₂O
MW: 235.03 g/mol
Melting Pt: 34…35 °C
MDL Number: MFCD00085010
CAS Number: 40706-98-7

Order Now

Specification Test Results

Melting point 34-35°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR