Potassium hexafluorophosphate ≥99%

Supplier: Matrix Scientific

001037-100G 001037-500G
101775-162EA 30.77 USD
101775-162 101775-164
Potassium hexafluorophosphate ≥99%
Potassium hexafluorophosphate

TCA listed: T
Hazard: Corrosive/Irritant/Hygroscopic

Formula: F₆KP
MW: 184.06 g/mol
Melting Pt: 575
Density: 2.55
MDL Number: MFCD00011412
CAS Number: 17084-13-8
UN: 3260
ADR: 8 II

Order Now

Specification Test Results

Melting point 575°
Density 2.55


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR