Silver fluoride ≥99%
Supplier: Matrix Scientific
Synonyms:
Silver monofluoride, Silver(I) fluoride, Argentous fluoride
001028-5G
001028-25G
001028-100G
101775-150EA
130.77
USD
101775-150
101775-152
101775-154
Silver fluoride ≥99%
Silver fluoride
TCA listed: T
Hazard: Corrosive/Stored under Argon/Light Sensitive/Hygroscopic
Formula:
AgF MW: 126.87 g/mol Boiling Pt: 1159 °C Melting Pt: 435 °C Density: 5.852 |
MDL Number:
MFCD00003410 CAS Number: 7775-41-9 UN: 3260 ADR: 8,II |
Specification Test Results
Boiling point | 1159° |
Density | 5.852 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...