Pyrazine ≥99%

Supplier: Matrix Scientific

001010-100G 001010-25G
101775-140EA 72.31 USD
101775-140 101775-138
Pyrazine ≥99%
Pyrazine

TCA listed: T
Hazard: Flammable Solid

Formula: C₄H₄N₂
MW: 80.09 g/mol
Boiling Pt: 115-116°C
Melting Pt: 54-56°C
Density: 1.031
MDL Number: MFCD00006122
CAS Number: 290-37-9
UN: 1325
ADR: 4.1,III

Order Now

Specification Test Results

Boiling point 115-116°
Density 1.031

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR