2,3,4-Trifluorobenzonitrile ≥98%

Supplier: Matrix Scientific

003670-5G 003670-25G
101776-976EA 21.54 USD
101776-976 101776-978
2,3,4-Trifluorobenzonitrile ≥98%
2,3,4-Trifluorobenzonitrile

Hazard: Toxic/Irritant

Formula: C₇H₂F₃N
MW: 157.09 g/mol
Boiling Pt: 116-118°C/100mm
Density: 1.12
MDL Number: MFCD00013288
CAS Number: 143879-80-5
UN: 3276
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 116-118°/1
Density 1.12
ND20 1.47

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR