Methyl-2-fluorobenzoate ≥98%
Supplier: Matrix Scientific
Synonyms:
Methyl 2-fluorobenzoate, 2-Fluorobenzoic acid emthyl ester
003669-25G
003669-100G
101776-972EA
27.69
USD
101776-972
101776-974
Methyl-2-fluorobenzoate ≥98%
Methyl-2-fluorobenzoate
TCA listed: T
Hazard: Flammable
Formula:
C₈H₇FO₂ MW: 154.14 g/mol Boiling Pt: 109-110°C/35mm Density: 1.19 |
MDL Number:
MFCD00017913 CAS Number: 394-35-4 |
Specification Test Results
Boiling point | 109-110°/3 |
Density | 1.19 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...