2,4,5-Trifluorobenzenesulfonyl chloride ≥97%

Supplier: Matrix Scientific

Synonyms: 2,4,5-Trifluorobenzene-1-sulphonyl chloride

003665-1G 003665-25G 003665-5G
101776-962EA 13.85 USD
101776-962 101776-966 101776-964
2,4,5-Trifluorobenzenesulfonyl chloride ≥97%
2,4,5-Trifluorobenzenesulphonyl chloride

Hazard: Corrosive/Irritant/Refrigerate

Formula: C₆H₂ClF₃O₂S
MW: 230.59 g/mol
Density: 1.657
MDL Number: MFCD00077600
CAS Number: 220227-21-4
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Density 1.657
ND20 1.503

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR