2,3,5-Trifluorobenzyl bromide

Supplier: Matrix Scientific

Synonyms: α-Bromo-2,3,5-trifluorotoluene

003662-1G 003662-5G
101776-954EA 44.62 USD
101776-954 101776-956
2,3,5-Trifluorobenzyl bromide
2,3,5-Trifluorobenzyl bromide

Hazard: Corrosive/Refrigerate/Lachrymator

Formula: C₇H₄BrF₃
MW: 225.01 g/mol
Boiling Pt: 182
Melting Pt: 21
MDL Number: MFCD00083543
CAS Number: 226717-83-5
UN: 3265
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR