2,3,5-Trifluorobenzoyl chloride

Supplier: Matrix Scientific

003655-1G 003655-5G
101776-950EA 63.08 USD
101776-950 101776-952
2,3,5-Trifluorobenzoyl chloride
2,3,5-Trifluorobenzoyl chloride

Hazard: Corrosive/Lachrymator

Formula: C₇H₂ClF₃O
MW: 194.54 g/mol
CAS Number: 240800-48-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR