2,3,5-Trifluorobenzonitrile

Supplier: Matrix Scientific

003654-1G 003654-5G
101776-946EA 40 USD
101776-946 101776-948
2,3,5-Trifluorobenzonitrile
2,3,5-Trifluorobenzonitrile

Hazard: Toxic

Formula: C₇H₂F₃N
MW: 157.09 g/mol
MDL Number: MFCD00083539
CAS Number: 241154-09-6
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR