2,4,6-Trifluorobenzyl bromide ≥97%

Supplier: Matrix Scientific

Synonyms: α-Bromo-2,4,6-trifluorotoluene, 2-(Bromomethyl)-1,3,5-trifluorobenzene

003650-1G 003650-5G
101776-938EA 175.38 USD
101776-938 101776-940
2,4,6-Trifluorobenzyl bromide ≥97%
2,4,6-Trifluorobenzyl bromide

TCA listed: T
Hazard: Corrosive

Formula: C₇H₄BrF₃
MW: 225.01 g/mol
MDL Number: MFCD00061210
CAS Number: 151411-98-2
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR