2,4,5-Trifluorobenzyl bromide ≥97%

Supplier: Matrix Scientific

Synonyms: 1-(Bromomethyl)-2,4,5-trifluorobenzene, α-Bromo-2,4,5-trifluorotoluene

003649-5G 003649-25G 003649-100G
101776-932EA 30.77 USD
101776-932 101776-934 101776-936
2,4,5-Trifluorobenzyl bromide ≥97%
2,4,5-Trifluorobenzyl bromide

TCA listed: T
Hazard: Corrosive

Formula: C₇H₄BrF₃
MW: 225.01 g/mol
MDL Number: MFCD00061209
CAS Number: 157911-56-3
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR