2,3,6-Trifluorobenzyl bromide ≥97%

Supplier: Matrix Scientific

003648-1G 003648-5G
101776-928EA 141.54 USD
101776-928 101776-930
2,3,6-Trifluorobenzyl bromide ≥97%
2,3,6-Trifluorobenzyl bromide

Hazard: Corrosive

Formula: C₇H₄BrF₃
Boiling Pt: 184°C (dec)
Density: 1.718
Flash Pt: 90°C
MDL Number: MFCD00061208
CAS Number: 151412-02-1
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Melting point 114°(dec)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR