2,3,4-Trifluorobenzyl bromide ≥97%

Supplier: Matrix Scientific

Synonyms: 1-(Bromomethyl)-2,3,4-trifluorobenzene, α-Bromo-2,3,4-trifluorotoluene

003647-5G 003647-1G
101776-926EA 115.38 USD
101776-926 101776-924
2,3,4-Trifluorobenzyl bromide ≥97%
2,3,4-Trifluorobenzyl bromide

TCA listed: T
Hazard: Corrosive

Formula: C₇H₄BrF₃
MW: 225.01 g/mol
Density: 1.71
Flash Pt: 90°C
MDL Number: MFCD00061233
CAS Number: 157911-55-2
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR