2,4,5-Trifluorobenzaldehyde ≥97%
Supplier: Matrix Scientific
003627-100G
003627-25G
101776-906EA
638.46
USD
101776-906
101776-904
2,4,5-Trifluorobenzaldehyde ≥97%
Hazard: Flammable
Formula:
C₇H₃F₃O Boiling Pt: 174 °C Density: 1.408 Flash Pt: 58 °C |
MDL Number:
MFCD00061196 CAS Number: 165047-24-5 UN: 1989 ADR: 3,III |
Specification Test Results
ND20 | 1.482 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...