2,4,5-Trifluorobenzoyl chloride ≥97%

Supplier: Matrix Scientific

Synonyms: 2,4,5-trifluorobenzoylchloride

003623-5G 003623-25G
101776-900EA 23.08 USD
101776-900 101776-902
2,4,5-Trifluorobenzoyl chloride ≥97%
2,4,5-Trifluorobenzoyl chloride

Hazard: Corrosive/Moisture Sensitive/Lachrymator

Formula: C₇H₂ClF₃O
MW: 194.54 g/mol
Density: 1.52
MDL Number: MFCD00061204
CAS Number: 88419-56-1
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Density 1.52
ND20 1.498

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR