2,3,5-Trifluoroaniline ≥97%

Supplier: Matrix Scientific

003612-5G 003612-25G
101776-882EA 227.69 USD
101776-882 101776-884
2,3,5-Trifluoroaniline ≥97%
2,3,5-Trifluoroaniline

Hazard: Toxic

Formula: C₆H₄F₃N
MW: 147.1 g/mol
MDL Number: MFCD00083549
CAS Number: 363-80-4
UN: 2941
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR