2,4,5-Trifluoroaniline ≥97%

Supplier: Matrix Scientific

003611-25G 003611-100G
101776-878EA 29.23 USD
101776-878 101776-880
2,4,5-Trifluoroaniline ≥97%
2,4,5-Trifluoroaniline

Hazard: Toxic/Irritant

Formula: C₆H₄F₃N
MW: 147.1 g/mol
Melting Pt: 59-60°C
MDL Number: MFCD00007649
CAS Number: 367-34-0
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 59-60°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR