Methyl difluoroacetate ≥99%

Supplier: Matrix Scientific

003610-25G 003610-100G
101776-874EA 49.23 USD
101776-874 101776-876
Methyl difluoroacetate ≥99%
Methyl difluoroacetate

Hazard: Flammable

Formula: C₃H₄F₂O₂
MW: 110.06 g/mol
Boiling Pt: 85…86 °C
Density: 1.272
MDL Number: MFCD00039276
CAS Number: 433-53-4
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Boiling point 85-86°
Density 1.272
ND20 1.337

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR