4,4,4-Trifluoro-3-(trifluoromethyl)crotonic acid

Supplier: Matrix Scientific

003581-5G 003581-1G
101776-848EA 149.23 USD
101776-848 101776-846
4,4,4-Trifluoro-3-(trifluoromethyl)crotonic acid
4,4,4-Trifluoro-3-(trifluoromethyl)crotonic acid

Hazard: Toxic/Irritant

Formula: C₅H₂F₆O₂
Boiling Pt: 90°C/51 Torr
MDL Number: MFCD00013574
CAS Number: 1763-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR