1,1,1-Trifluoro-2-propanol ≥97%

Supplier: Matrix Scientific

003575-5G 003575-25G
101776-842EA 192.31 USD
101776-842 101776-844
1,1,1-Trifluoro-2-propanol ≥97%
1,1,1-Trifluoro-2-propanol

TCA listed: T
Hazard: Flammable

Formula: C₃H₅F₃O
Boiling Pt: 75-76°C
MDL Number: MFCD00014403
CAS Number: 374-01-6
UN: 1987
ADR: 3,III

Order Now

Specification Test Results

Boiling point 75-76°
ND20 1.3254


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR