1,1,1-Trifluoro-2-butanone cyanohydrin ≥95%

Supplier: Matrix Scientific

003570-1G 003570-5G
101776-838EA 33.85 USD
101776-838 101776-840
1,1,1-Trifluoro-2-butanone cyanohydrin ≥95%
1,1,1-Trifluoro-2-butanone cyanohydrin

Hazard: Toxic/Irritant

Formula: C₅H₆F₃NO
Boiling Pt: 75-78°C/40mm
MDL Number: MFCD00236720
CAS Number: 203302-91-4

Order Now

Specification Test Results

Boiling point 75-78°/40m

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR