1,1,1-Trifluoro-2-butene ≥94%

Supplier: Matrix Scientific

003568-5G 003568-25G
101776-830EA 130.77 USD
101776-830 101776-832
1,1,1-Trifluoro-2-butene ≥94%
1,1,1-Trifluoro-2-butene

Hazard: Flammable Gas/Store in Freezer

Formula: C₄H₅F₃
Boiling Pt: 17 °C
MDL Number: MFCD00082625
CAS Number: 406-39-3

Order Now

Specification Test Results

Boiling point 17°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR