1,1,1-Trifluoroacetylacetone ≥98%
Supplier: Matrix Scientific
Synonyms:
1,1,1-Trifluoropentane-2,4-dione, 1,1,1-Trifluoro-2,4-pentanedione
003565-25G
003565-100G
101776-826EA
95.38
USD
101776-826
101776-828
1,1,1-Trifluoroacetylacetone ≥98%
1,1,1-Trifluoroacetylacetone
TCA listed: T
Hazard: Flammable/Refrigerate
Formula:
C₅H₅F₃O₂ MW: 154.09 g/mol Boiling Pt: 105-107°C Density: 1.27 |
MDL Number:
MFCD00000427 CAS Number: 367-57-7 UN: 1224 ADR: 3,III |
Specification Test Results
Boiling point | 105-107° |
Density | 1.27 |
ND20 | 1.389 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...