1H,1H-Heptacosafluoro-1-tetradecanol ≥95%

Supplier: Matrix Scientific

Synonyms: 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,14-Heptacosafluoro-1-tetradecanol, 1H,1H-Perfluoro-1-tetradecanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,14-Heptacosafluor-1-tetradecanol

003562-5G 003562-25G
101776-814EA 52.31 USD
101776-814 101776-816
1H,1H-Heptacosafluoro-1-tetradecanol ≥95%
1H,1H-Heptacosafluoro-1-tetradecanol

Hazard: Irritant

Formula: C₁₄H₃F₂₇O
MW: 700.13 g/mol
Boiling Pt: 130°C/20mm
Melting Pt: 129-132°C
MDL Number: MFCD00153264
CAS Number: 15622-57-8

Order Now

Specification Test Results

Boiling point 130°/20mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR