4,4,4-Trifluoro-1-butanol ≥98%
Supplier: Matrix Scientific
003553-25G
003553-5G
101776-812EA
243.08
USD
101776-812
101776-810
4,4,4-Trifluoro-1-butanol ≥98%
4,4,4-Trifluoro-1-butanol
Hazard: Flammable
Formula:
C₄H₇F₃O Boiling Pt: 123…125 °C |
MDL Number:
MFCD00041483 CAS Number: 461-18-7 UN: 1993 ADR: 3,III |
Specification Test Results
Boiling point | 123-125° |
ND20 | 1.3425 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...