1,1,1,3-Tetrafluoro-2-(trifluoromethyl)-4-oxapent-2-ene ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 1,3,3,3-tetrafluoro-2-(trifluoromethyl)prop-1-en-1-yl ether

003511-5G 003511-25G
101776-794EA 661.54 USD
101776-794 101776-796
1,1,1,3-Tetrafluoro-2-(trifluoromethyl)-4-oxapent-2-ene ≥97%
1,1,1,3-Tetrafluoro-2-(trifluoromethyl)-4-oxapent-2-ene

Hazard: Flammable

Formula: C₅H₃F₇O
MW: 212.07 g/mol
Boiling Pt: 98-104°C
Density: 1.508
MDL Number: MFCD00054679
CAS Number: 360-53-2

Order Now

Specification Test Results

Boiling point 98-104°
Density 1.508
ND20 1.317

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR