1,2,3,4-Tetrafluorobenzene ≥99%

Supplier: Matrix Scientific

003510-1G 003510-5G
101776-788EA 15.38 USD
101776-788 101776-790
1,2,3,4-Tetrafluorobenzene ≥99%
1,2,3,4-Tetrafluorobenzene

Hazard: Flammable/Refrigerate

Formula: C₆H₂F₄
MW: 150.08 g/mol
Boiling Pt: 95°C
Melting Pt: -42°C
Density: 1.4
MDL Number: MFCD00000285
CAS Number: 551-62-2
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Boiling point 95°
Density 1.4
ND20 1.408

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR