1,2,4,5-Tetrafluorobenzene ≥99%
Supplier: Matrix Scientific
003507-25G
003507-100G
101776-784EA
84.62
USD
101776-784
101776-786
1,2,4,5-Tetrafluorobenzene ≥99%
1,2,4,5-Tetrafluorobenzene
Hazard: Flammable
Formula:
C₆H₂F₄ MW: 150.08 g/mol Boiling Pt: 90 °C Melting Pt: 4 °C Density: 1.344 |
MDL Number:
MFCD00000307 CAS Number: 327-54-8 UN: 2387 ADR: 3,II |
Specification Test Results
Boiling point | 90° |
Density | 1.344 |
ND20 | 1.407 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...