2,3,5,6-Tetrafluorophenol, Technical Grade
Supplier: Matrix Scientific
003484-5G
003484-25G
101776-756EA
21.54
USD
101776-756
101776-758
2,3,5,6-Tetrafluorophenol, Technical Grade
2,3,5,6-Tetrafluorophenol
Hazard: Corrosive
Formula:
C₆H₂F₄O Boiling Pt: 140 °C Melting Pt: 37…39 °C |
MDL Number:
MFCD00002157 CAS Number: 769-39-1 UN: 3261 ADR: 8,II |
Specification Test Results
Boiling point | 140° |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...